Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Profilin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7779
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7779 Supplier R&D Systems Supplier No. MAB7779
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Profilin 1 Monoclonal specifically detects Profilin 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Profilin 1
Applications Western Blot
Classification Monoclonal
Clone 816536
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P07737
Gene Alias PFN1, PROF1, profilin 1, Profilin I, profilin-1
Gene Symbols PFN1
Host Species Mouse
Immunogen E. coli-derived recombinant human Profilin 1 Met1-Tyr140 Accession # P07737
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5216
Test Specificity Detects human Profilin 1 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Profilin 2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.