Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Progesterone R B/NR3C3 B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH534400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH534400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH534400 Supplier R&D Systems Supplier No. PPH534400
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Progesterone R B/NR3C3 Monoclonal specifically detects Progesterone R B/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.

Specifications

Antigen Progesterone R B/NR3C3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA
Classification Monoclonal
Clone H5344
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation, Direct ELISA
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Symbols PGR
Host Species Mouse
Immunogen Recombinant human Progesterone R B/NR3C3 aa 2-99
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specifically reacts with human PR isoform B and cross-reacts with mouse and rat PR isoform B. It does not cross-react with human PR isoform A. Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.