Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Progranulin/PGRN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Progranulin/PGRN |
---|---|
Clone | 296612 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24202SP
![]() |
R&D Systems
MAB24202SP |
25 μg |
Each for $200.50
|
|
|||||
MAB24202100
![]() |
R&D Systems
MAB24202100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Human samples. It is validated for ELISA.Specifications
Progranulin/PGRN | |
ELISA | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
Mouse myeloma cell line NS0-derived recombinant human Progranulin/PGRN, Met1-Leu593, Accession # P28799 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Progranulin/PGRN in direct ELISAs. |
296612 | |
ELISA | |
Unconjugated | |
Human | |
P28799 | |
2896 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title