Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Prokineticin R1/PROKR1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4655G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4655G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4655G100 Supplier R&D Systems Supplier No. FAB4655G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prokineticin R1/PROKR1 Monoclonal specifically detects Prokineticin R1/PROKR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Prokineticin R1/PROKR1
Applications Flow Cytometry
Classification Monoclonal
Clone 420849
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8TCW9
Gene Alias G protein-coupled receptor 73, G protein-coupled receptor ZAQ, GPR73, GPR73a, GPR73aG-protein coupled receptor 73, PKR1, PK-R1, PKR1G-protein coupled receptor ZAQ, prokineticin receptor 1, ProkineticinR1, PROKR1, ZAQ
Gene Symbols PROKR1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Prokineticin R1/PKR1 Met1-Lys393 Accession # Q8TCW9
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10887
Test Specificity Detects human Prokineticin R1/PKR1. Stains human Prokineticin R1/PKR1 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.