Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Prolactin R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1167
This item is not returnable.
View return policy
Description
Prolactin R Monoclonal specifically detects Prolactin R in Human samples. It is validated for Western Blot, Neutralization.Specifications
Prolactin R | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 2.5-9 ug/mL | |
P16471 | |
PRLR | |
Mouse myeloma cell line NS0-derived recombinant human Prolactin R Gln25-Asp234 Accession # P16471 | |
500 μg | |
Primary | |
Detects human Prolactin R in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Prolactin R and recombinant rat Prolactin R is observed. | |
Human | |
Purified |
Western Blot, Neutralization | |
250448 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRLR, PRL-R, prolactin receptor, prolactin receptor delta 7/11, ProlactinR, secreted prolactin binding protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5618 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction