Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Prominin 2 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2024G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2024G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2024G100 Supplier R&D Systems Supplier No. FAB2024G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prominin 2 Monoclonal specifically detects Prominin 2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Prominin 2
Applications Flow Cytometry
Classification Monoclonal
Clone 244029
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAM10541
Gene Alias hPROML2, MGC138714, PROM2, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2
Gene Symbols PROM2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Prominin 2 Ala27-Leu834 Accession # AAM10541
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150696
Test Specificity Detects human Prominin 2. Stains human Prominin 2-transfected cells but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.