Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Prominin 2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2024S100UG
This item is not returnable.
View return policy
Description
Prominin 2 Monoclonal specifically detects Prominin 2 in Human samples. It is validated for Flow Cytometry.Specifications
Prominin 2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAM10541 | |
PROM2 | |
NS0 mouse myeloma cell line transfected with human Prominin 2 Ala27-Leu834 Accession # AAM10541 | |
RUO | |
150696 | |
Human | |
IgG2b |
Flow Cytometry | |
244029 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
hPROML2, MGC138714, PROM2, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2 | |
Mouse | |
100 μg | |
Primary | |
Detects human Prominin 2. Stains human Prominin 2-transfected cells but not irrelevant transfectants. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction