Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB38881
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Proprotein Convertase 9/PCSK9 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8NBP7 | |
| PCSK9 | |
| Mouse myeloma cell line NS0-derived recombinant human Proprotein Convertase 9/PCSK9 isoform 1 Arg29-Gln692 Accession # Q8NBP7 | |
| 100 μg | |
| Primary | |
| Detects the mature form of human Proprotein Convertase 9/PCSK9 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) PCSK1, rhPCSK7, or recombinant mouse PCSK9. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 410420 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 255738 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction