Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.50
Specifications
Antigen | Proprotein Convertase 9/PCSK9 |
---|---|
Clone | 499911 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB38882SP
![]() |
R&D Systems
MAB38882SP |
25 μg |
Each for $200.50
|
|
|||||
MAB38882
![]() |
R&D Systems
MAB38882 |
100 μg |
Each for $468.50
|
|
|||||
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Proprotein Convertase 9/PCSK9 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Human | |
Q8NBP7 | |
255738 | |
Mouse myeloma cell line NS0-derived recombinant human Proprotein Convertase 9/PCSK9 Arg29-Gln692 Accession # Q8NBP7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Proprotein Convertase 9/PCSK9 in direct ELISAs and Western blots. In Western blots, 10% cross-reactivity with recombinant mouse PCSK9 is observed and no cross-reactivity with recombinant human (rh) PCSK1 or rhPCSK7 is observed. |
499911 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
PCSK9 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title