Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF3888
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Polyclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Proprotein Convertase 9/PCSK9 | |
Polyclonal | |
Unconjugated | |
Q8NBP7 | |
PCSK9 | |
Mouse myeloma cell line NS0-derived recombinant human PCSK9 isoform 1 Gln31-Gln692 Accession # Q8NBP7 | |
100 μg | |
Primary | |
Detects human PCSK9 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse PCSK9 is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
Sheep | |
Affinity Purified | |
RUO | |
255738 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction