Learn More
Description
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 125.00 to 8000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 255738 |
| Product Line | R&D Systems |
| Synonym | EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
