Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Proprotein Convertase 9/PCSK9 (Research Grade Alirocumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
$188.28 - $543.90
Specifications
| Antigen | Proprotein Convertase 9/PCSK9 |
|---|---|
| Clone | Hu135 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10347SP
![]() |
R&D Systems
MAB10347SP |
25 μg |
Each for $188.28
|
|
|||||
MAB10347100
![]() |
R&D Systems
MAB10347100 |
100 μg |
Each for $543.90
|
|
|||||
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| Proprotein Convertase 9/PCSK9 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 255738 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| Hu135 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Human | |
| Human | |
| EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Human Proprotein Convertase 9/PCSK9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detccts Human Proprotein Convertase 9/PCSK9 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alirocumab. This product is for research use only. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title