Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Prostaglandin E Synthase 2/PTGES2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7627100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7627100 100 μg
MAB7627SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7627100 Supplier R&D Systems Supplier No. MAB7627100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prostaglandin E Synthase 2/PTGES2 Monoclonal specifically detects Prostaglandin E Synthase 2/PTGES2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Prostaglandin E Synthase 2/PTGES2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 998012
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9H7Z7
Gene Alias C9orf15, chromosome 9 open reading frame 15, EC 5.3.99.3, FLJ14038, gamma-interferon-activated transcriptional element-binding factor 1, GATE-binding factor 1, GBF1, membrane-associated prostaglandin E synthase 2, Microsomal prostaglandin E synthase 2, microsomal prostaglandin E synthase-2, mPGES2, mPGES-2, PGES2, PGES2MGC11289, prostaglandin E synthase 2
Host Species Mouse
Immunogen E. coli-derived recombinant Human Prostaglandin E Synthase 2/PTGES2, Glu88-His377, Accession No. Q9H7Z7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 80142.0
Test Specificity Detects Human Prostaglandin E Synthase 2/PTGES2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.