Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Prostaglandin I2 Synthase Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7788SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7788SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7788SP Supplier R&D Systems Supplier No. MAB7788SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prostaglandin I2 Synthase Monoclonal specifically detects Prostaglandin I2 Synthase in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Prostaglandin I2 Synthase
Applications Western Blot
Classification Monoclonal
Clone 852307
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16647
Gene Alias CYP8, CYP8A1, CYP8A1PTGI, CYP8prostacyclin synthase, EC 5.3.99.4, MGC126858, MGC126860, PGIS, PGIScytochrome P450, family 8, subfamily A, polypeptide 1, Prostacyclin Synthase, prostaglandin I2 (prostacyclin) synthase, Prostaglandin I2 synthase, PTGI
Gene Symbols PTGIS
Host Species Mouse
Immunogen E. coli-derived recombinant human Prostaglandin I2 Synthase Arg21-Pro500 Accession # Q16647
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5740
Test Specificity Detects human Prostaglandin I2 Synthase in ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.