Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Prostatic Acid Phosphatase/ACPP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $526.50
Specifications
Antigen | ProstaticAcidPhosphatase/ACPP |
---|---|
Clone | 911603R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB62401SP
![]() |
R&D Systems
MAB62401SP |
25 μg |
Each for $241.00
|
|
|||||
MAB62401100
![]() |
R&D Systems
MAB62401100 |
100 μg |
Each for $526.50
|
|
|||||
Description
Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for ELISA.Specifications
ProstaticAcidPhosphatase/ACPP | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P15309 | |
55 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
911603R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
acid phosphatase, prostate, ACP3, ACP-3, ACPP, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
Mouse myeloma cell line NS0-derived recombinant human Prostatic Acid Phosphatase/ACPP Met1-Gln379 Accession # P15309 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Prostatic Acid Phosphatase/ACPP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title