Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Prostatic Acid Phosphatase/ACPP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6240
This item is not returnable.
View return policy
Description
Prostatic Acid Phosphatase/ACPP Polyclonal specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Prostatic Acid Phosphatase/ACPP | |
Polyclonal | |
Unconjugated | |
P15309 | |
ACPP | |
Mouse myeloma cell line NS0-derived recombinant human Prostatic Acid Phosphatase/ACPP Lys33-Gln379 Accession # P15309 | |
100 μg | |
Primary | |
Detects human Prostatic Acid Phosphatase/ACPP in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
acid phosphatase, prostate, ACP3, ACP-3, ACPP, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
Sheep | |
Affinity Purified | |
RUO | |
55 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction