Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Protein Disulfide Isomerase/P4HB Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4236G
This item is not returnable.
View return policy
Description
Protein Disulfide Isomerase/P4HB Monoclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Immunocytochemistry.Specifications
Protein Disulfide Isomerase/P4HB | |
Monoclonal | |
Alexa Fluor 488 | |
P07237 | |
P4HB | |
Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB Asp18-Lys505 Accession # P07237 | |
100 Tests | |
5034 | |
Human | |
Purified |
Immunocytochemistry | |
537331 | |
Immunocytochemistry 1:10 dilution | |
Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDI, PDIA1, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHB, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, Thbp, thyroid hormone-binding protein p55 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Conjugated Protein Disulfide Isomerase/P4HB antibodies are ideal for immunocytochemistry colocalization studies in the lumen of the endoplasmic reticulum. The unconjugated antibody detects human Protein Disulfide Isomerase/P4HB in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction