Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Proteinase 3/Myeloblastin/PRTN3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6134 Supplier R&D Systems Supplier No. MAB6134
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Proteinase 3/Myeloblastin/PRTN3
Applications Western Blot
Classification Monoclonal
Clone 684042
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P24158
Gene Alias ACPA, AGP7, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, PRTN3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen
Gene Symbols PRTN3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf21-derived recombinant human Proteinase 3/Myeloblastin/PRTN3 Ala26-Arg249 Accession # P24158
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5657
Test Specificity Detects human Proteinase 3/Myeloblastin/PRTN3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Granzyme B is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.