Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Proteinase 3/Myeloblastin/PRTN3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB61341
This item is not returnable.
View return policy
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Immunocytochemistry.Specifications
Proteinase 3/Myeloblastin/PRTN3 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P24158 | |
PRTN3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Proteinase 3/Meyeloblastin/PRTN3 Ala26-Arg249 Accession # P24158 | |
100 μg | |
Primary | |
Detects human Proteinase 3/Meyeloblastin/PRTN3 in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
684022 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACPA, AGP7, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, PRTN3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5657 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction