Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Protocadherin-15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6729SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6729SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6729SP Supplier R&D Systems Supplier No. MAB6729SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protocadherin-15 Monoclonal specifically detects Protocadherin-15 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Protocadherin-15
Applications Immunocytochemistry
Classification Monoclonal
Clone 740523
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96QU1
Gene Alias autosomal recessive 23, cadherin-related family member 15, CDHR15, DFNB23, DKFZp667A1711, PCDH15, protocadherin-15, Protocadherin15, protocadherin-related 15, USH1F
Gene Symbols PCDH15
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin-15 Tyr28-Ala1376 Accession # Q96QU1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65217
Test Specificity Detects human Protocadherin-15 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.