Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Protocadherin gamma C3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8364
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8364 100 μg
MAB8364SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8364 Supplier R&D Systems Supplier No. MAB8364
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protocadherin gamma C3 Monoclonal specifically detects Protocadherin gamma C3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Protocadherin gamma C3
Applications Western Blot
Classification Monoclonal
Clone 926525
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UN70
Gene Alias cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, PCDHGC3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43
Gene Symbols PCDHGC3
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3 Met1-Tyr693 Accession # Q9UN70
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5098
Test Specificity Detects human Protocadherin gamma C3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.