Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Protocadherin gamma C3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Protocadherin gamma C3 |
---|---|
Clone | 926518 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB83641SP
![]() |
R&D Systems
MAB83641SP |
25 μg |
Each for $200.50
|
|
|||||
MAB83641100
![]() |
R&D Systems
MAB83641100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Protocadherin gamma C3 Monoclonal specifically detects Protocadherin gamma C3 in Human samples. It is validated for Flow Cytometry.Specifications
Protocadherin gamma C3 | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Mouse | |
Human | |
Q9UN70 | |
5098 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
926518 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, PCDHGC3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3 Met1-Tyr693 Accession # Q9UN70 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Protocadherin gamma C3 in direct ELISAs. Stains human Protocadherin gamma C3 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title