Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Protogenin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6795SP
This item is not returnable.
View return policy
Description
Protogenin Polyclonal specifically detects Protogenin in Human samples. It is validated for Immunohistochemistry.Specifications
Protogenin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), PRTG, RPTG, Shen-Dan | |
Sheep | |
Affinity Purified | |
RUO | |
283659 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
Q2VWP7 | |
PRTG | |
Mouse myeloma cell line NS0-derived recombinant human Protegenin Phe36-Ala952 Accession # Q2VWP7 | |
25 μg | |
Primary | |
Detects human Protegenin in direct ELISAs. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse Protegenin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction