Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Protogenin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6795
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6795 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6795 Supplier R&D Systems Supplier No. AF6795
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Protogenin Polyclonal specifically detects Protogenin in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Protogenin
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q2VWP7
Gene Alias FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), PRTG, RPTG, Shen-Dan
Gene Symbols PRTG
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Protegenin Phe36-Ala952 Accession # Q2VWP7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283659
Test Specificity Detects human Protegenin in direct ELISAs. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse Protegenin is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.