Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PSG6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB85983100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB85983100 100 μg
MAB85983SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB85983100 Supplier R&D Systems Supplier No. MAB85983100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSG6 Monoclonal specifically detects PSG6 in Human samples. It is validated for ELISA.

Specifications

Antigen PSG6
Applications ELISA
Classification Monoclonal
Clone 935947
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q00889
Gene Alias CGM3, pregnancy specific beta-1-glycoprotein 6, Pregnancy-specific beta-1-glycoprotein 10, Pregnancy-specific beta-1-glycoprotein 12, pregnancy-specific beta-1-glycoprotein 6, Pregnancy-specific glycoprotein 10, Pregnancy-specific glycoprotein 12, Pregnancy-specific glycoprotein 6, PS-beta-G-10, PS-beta-G-12, PS-beta-G-6, PSBG-10, PSBG-12, PSBG-6, PSG10, PSG12, PSG-6, PSGGB
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK-293-derived transfected with human PSG6 Met1-Gly424 Accession # Q00889
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5675
Test Specificity Detects human PSG6 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.