Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PSGL-1/CD162 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB996
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB996 500 μg
MAB996SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB996 Supplier R&D Systems Supplier No. MAB996
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Adhesion Blockade, CyTOF-ready.

Specifications

Antigen PSGL-1/CD162
Applications Flow Cytometry, Adhesion Blockade, CyTOF
Classification Monoclonal
Clone CHO131
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG
Gene Symbols SELPLG
Host Species Mouse
Immunogen CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6404
Test Specificity Detects human PSGL-1/CD162. Recognizes sLe x -bearing core 2 O-glycan stuctures. It does not recognize sLe x on an extended core 1 O-glycan. The sLe x -bearing, core 2 O-glycan structure decorates the P-Selectin ligand PSGL-1, and the presence of this glycan structure is required for high affinity P-Selectin binding (1). This antibody stains human and canine leukocytes but does not recognize monkey, mouse, rabbit, porcine, feline or bovine leukocytes.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.