Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PSGL-1/CD162 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB9961SP
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
PSGL-1/CD162 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
NP_002997 | |
SELPLG | |
Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1/CD162 Gln42-Gly295 Accession # NP_002997 | |
25 μg | |
Primary | |
Detects human PSGL-1/CD162 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
688101 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6404 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction