Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PSMA/FOLH1/NAALADase I Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4234G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4234G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4234G Supplier R&D Systems Supplier No. FAB4234G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSMA/FOLH1/NAALADase I Monoclonal specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PSMA/FOLH1/NAALADase I
Applications Flow Cytometry
Classification Monoclonal
Clone 460420
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q04609
Gene Alias Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PSMA/FOLH1/NAALADase I Lys44-Ala750 Accession # Q04609
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2346
Test Specificity Detects human PSMA/FOLH1/NAALADase I in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) NAALADase-like 2 and no cross-reactivity with rhNAALADase-like 1, rhNAALADase-like 3, recombinant mouse (rm) PSMA/FOLH1/NAALADase I, or rmNAALAase-like 2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.