Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PSMA/FOLH1/NAALADase I Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4234
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4234 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4234 Supplier R&D Systems Supplier No. AF4234
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PSMA/FOLH1/NAALADase I Polyclonal specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen PSMA/FOLH1/NAALADase I
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, ELISA, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. Q04609
Gene Alias Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PSMA/FOLH1/NAALADase I Lys44-Ala750 Accession # Q04609
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2346
Test Specificity Detects human PSMA/FOLH1/NAALADase I in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse NAALADase II is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.