Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PSMA/FOLH1 Phycoerythrin MAb (Clone 107-1A4), R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems K01425
This item is not returnable.
View return policy
Description
PSMA/FOLH1/NAALADase I Monoclonal specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Flow Cytometry.Specifications
PSMA/FOLH1/NAALADase I | |
Monoclonal | |
R-PE | |
PBS containing 1% BSA and 0.09% NaN 3 with 0.09% Sodium Azide | |
FOLH1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2346 | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
IgG1 |
Flow Cytometry | |
107-1A4 | |
Flow Cytometry | |
Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase | |
Mouse | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction