Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PTGER1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB100081SP
This item is not returnable.
View return policy
Description
PTGER1 Monoclonal specifically detects PTGER1 in Human samples. It is validated for Immunohistochemistry.Specifications
PTGER1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
EP1, PGE receptor EP1 subtype, PGE receptor, EP1 subtype, PGE2 receptor EP1 subtype, prostaglandin E receptor 1 (subtype EP1), 42kD, prostaglandin E receptor 1 (subtype EP1), 42kDa, prostaglandin E receptor 1, subtype EP1, prostaglandin E2 receptor EP1 subtype, Prostanoid EP1 receptor | |
Synthetic peptide containing human PTGER1, Accession # P34995 | |
25 μg | |
Primary | |
Detects human PTGER1 in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
1014918 | |
Immunohistochemistry 5-25 ug/mL | |
P34995 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5731 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction