Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PTGER2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6656SP
This item is not returnable.
View return policy
Description
PTGER2 Monoclonal specifically detects PTGER2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| PTGER2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P43116 | |
| PTGER2 | |
| NS0 mouse myeloma cell line transfected with human PTGER2 Accession # P43116 | |
| 25 μg | |
| Primary | |
| Detects human PTGER2 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 607077 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EP2, PGE receptor EP2 subtype, PGE2 receptor EP2 subtype, prostaglandin E receptor 2 (subtype EP2), 53kD, prostaglandin E receptor 2 (subtype EP2), 53kDa, prostaglandin E2 receptor EP2 subtype, Prostanoid EP2 receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5732 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction