Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PTGFR Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $467.50

Specifications

Antigen PTGFR
Clone 1014602
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10294SP
GREENER_CHOICE
View Documents
R&D Systems
MAB10294SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB10294100
GREENER_CHOICE
View Documents
R&D Systems
MAB10294100
100 μg
Each for $467.50
Only null left
Add to Cart
 
Description

Description

PTGFR Monoclonal specifically detects PTGFR in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

PTGFR
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
FP, MGC120498, MGC46203, PGF receptor, PGF2 alpha receptor, PGF2-alpha Receptor, prostaglandin F receptor (FP), prostaglandin F2 alpha receptor, prostaglandin F2-alpha receptor, prostaglandin receptor (2-alpha), Prostanoid FP receptor
Synthetic peptide containing human PTGFR RD3 epitope, Accession # P43088
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PTGFR in direct ELISAs.
1014602
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
P43088
5737
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.