Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PTH1R/PTHR1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5709
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5709 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5709 Supplier R&D Systems Supplier No. BAF5709
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PTH1R/PTHR1 Polyclonal specifically detects PTH1R/PTHR1 in Human samples. It is validated for Western Blot.

Specifications

Antigen PTH1R/PTHR1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q03431
Gene Alias MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHR, PTHR1, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor
Gene Symbols PTH1R
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PTH1R/PTHR1 Tyr23-Met189 Accession # Q03431
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5745
Test Specificity Detects human PTH1R/PTHR1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.