Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PTK2 (NP_722560.1, 1 a.a. - 527 a.a.) Full-length Recombinant Protein with GST-tag co-expressed with His-tagged WNK1 (NP_061852.1, 1 a.a. - 491 a.a.) expressed in Baculovirus infected Sf21 cells

Catalog No. 89958727
Click to view available options
Quantity:
5 μg

Used for Func, SDS-PAGE

This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined. [provided by RefSeq]

Specifications

Accession Number NP_722560.1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 5747
Molecular Weight (g/mol) 103kDa
Name PTK2 (Human) Recombinant Protein
Preparation Method Baculovirus infected insect cell (Sf21) expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FADK/FAK/FAK1/pp125FAK
Common Name PTK2
Gene Symbol PTK2
Biological Activity The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000.
Substrate : Blk/Lyntide. ATP: 100 μM.
Species Baculovirus
Recombinant Recombinant
Protein Tag GST
Expression System Baculovirus infected insect cell (Sf21) expression system
Form Liquid
Purity or Quality Grade 62% by SDS-PAGE/CBB staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.