Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PTP beta/zeta/PTPRZ Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB26881SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB26881SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB26881SP Supplier R&D Systems Supplier No. MAB26881SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PTP beta/zeta/PTPRZ Monoclonal specifically detects PTP beta/zeta/PTPRZ in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen PTP beta/zeta/PTPRZ
Applications Immunohistochemistry
Classification Monoclonal
Clone 279245
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23471
Gene Alias 3F8 CSPG, HPTPZ, phosphacan, protein tyrosine phosphatase, receptor-type, Z polypeptide 1, PTP18, PTPRZ, PTPRZ1, PTPZ, receptor-type, zeta polypeptide 1, RPTPB, RPTPbeta
Gene Symbols PTPRZ1
Host Species Rat
Immunogen E. coli-derived recombinant human PTP beta/zeta/PTPRZ Gly302-Asn754 Accession # P23471
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5803
Test Specificity Detects human PTP beta/zeta/PTPRZ in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.