Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PTP rho/PTPRT Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3697SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3697SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3697SP Supplier R&D Systems Supplier No. AF3697SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PTP rho/PTPRT Polyclonal specifically detects PTP rho/PTPRT in Human samples. It is validated for Western Blot.

Specifications

Antigen PTP rho/PTPRT
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O14522
Gene Alias EC 3.1.3, EC 3.1.3.48, KIAA0283RPTPrho, protein tyrosine phosphatase, receptor type, T, PTPRT, receptor protein tyrosine phosphatase, Receptor-type tyrosine-protein phosphatase rho, receptor-type tyrosine-protein phosphatase T, RPTP-rho, R-PTP-T
Gene Symbols PTPRT
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PTP rho/PTPRT Gln26-Lys725 Accession # O14522
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11122
Test Specificity Detects human PTP rho/PTPRT in Western blots. In Western blots, no cross-reactivity with recombinant human LAR, PTPRM, K, S, G or DEP1/CD148 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.