Learn More
Human PTP1B Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | PTP1B/PTPN1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Alias | EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, PTPN1, tyrosine-protein phosphatase non-receptor type 1 |
| Gene Symbols | PTPN1 |
| Host Species | Rabbit |
| Immunogen | E. coli-derived recombinant human PTP1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.