Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PU.1/Spi-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5870T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5870T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5870T100 Supplier R&D Systems Supplier No. IC5870T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PU.1/Spi-1 Monoclonal specifically detects PU.1/Spi-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen PU.1/Spi-1
Applications Flow Cytometry
Classification Monoclonal
Clone 732322
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_001074016
Gene Alias hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
Gene Symbols SPI1
Host Species Mouse
Immunogen E. coli-derived recombinant human PU.1/Spi-1 Met1-Lys169 Accession # NP_001074016
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6688
Test Specificity Detects human PU.1/Spi-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Spi-B, recombinant mouse (rm) PU.1/Spi-1, or rmSpi-B is observed. In Western blots, approximately 25% cross-reactivity with rhSpi-B and rmPU.1/Spi-1 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.