Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PU.1/Spi-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5870
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Monoclonal specifically detects PU.1/Spi-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
PU.1/Spi-1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_001074016 | |
SPI1 | |
E. coli-derived recombinant human PU.1/Spi-1 Met1-Lys169 Accession # NP_001074016 | |
100 μg | |
Primary | |
Detects human PU.1/Spi-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Spi-B, recombinant mouse (rm) PU.1/Spi-1, or rmSpi-B is observed. In Western blots, approximately 25% cross-reactivity with rhSpi-B and rmPU.1/Spi-1 is observed. | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
732322 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6688 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction