Learn More
Specifications
Specifications
| Assay Range | 3.13 to 200 ng/mL |
| Assay Sensitivity | 1.88 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human Pyridinoline in samples. No Significant cross-reactivity or interference between Human Pyridinoline and analogues was observed. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | (+)-pyridinoline, 1-(2 S)-(+)-pyridinoline, 4-((2 S)-2-amino-2-carboxyethyl)-1-((2 R,5 S)-5-amino-5-carboxy-2-hydroxypentyl)-3-((3 S)-3-amino-3-carboxypropyl)-5-hydroxy-Pyridinium inner salt, 4-[(2 S)-2-AMINO-2-CARBOXYETHYL]-3-[(3 S)-3-AMINO-3-CARBOXYPROPYL]-1-[(2 R,5 S)-5-AMINO-5-CARBOXYLATO-2-HYDROXYPENTYL]-5-HYDROXYPYRIDIN-1-IUM, Hydroxylysylpyridinoline, Pyridinium, 4-((2 S)-2-amino-2-carboxyethyl)-1-((2 R,5 S)-5-amino-5-carboxy-2-hydroxypentyl)-3-((3 S)-3-amino-3-carboxypropyl)-5-hydroxy-, inner salt |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
