Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Pyruvate Dehydrogenase E1-alpha subunit ELISA Kit (Colorimetric)
SDP

Catalog No. NB016779 Shop All R&D Systems Products
Change view
Click to view available options
Assay Range:
0.16 to 10 ng/mL
31.25 to 2000 pg/mL
Assay Sensitivity:
0.09 ng/mL
18.75 pg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Assay Range Assay Sensitivity
NB016779 0.16 to 10 ng/mL 0.09 ng/mL
NB016777 31.25 to 2000 pg/mL 18.75 pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB016779 Supplier Novus Biologicals™ Supplier No. NBP281220

This kit recognizes Human Pyruvate Dehydrogenase E1-alpha subunit in samples.

No significant cross-reactivity or interference between Human Pyruvate Dehydrogenase E1-alpha subunit and analogues was observed.

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 5160
Gene Symbol PDHA1
Host Species Human
Quantity 1 Kit
Research Discipline Cancer, Core ESC Like Genes, Lipid and Metabolism, Phospho Specific, Signal Transduction, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Pyruvate Dehydrogenase E1-alpha subunit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.