Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human R-Spondin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4645
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4645 100 μg
AF4645SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4645 Supplier R&D Systems Supplier No. AF4645
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

R-Spondin 1 Polyclonal specifically detects R-Spondin 1 in Human samples. It is validated for Neutralization.

Specifications

Antigen R-Spondin 1
Applications Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 3-12 ug/mL
Gene Accession No. Q2MKA7
Gene Alias Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Gene Symbols RSPO1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived human R-Spondin 1 Arg31-Ala263 Accession # Q2MKA7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284654
Test Specificity Detects human R-Spondin 1 in direct ELISAs. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse R-Spondin 1 is observed and less than 1% cross-reactivity with recombinant human (rh) R-Spondin 2, rhR-Spondin 3, and rhR-Spondin 4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.