Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human R-Spondin 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
| Antigen | R-Spondin 2 |
|---|---|
| Clone | 879709 |
| Concentration | LYOPH |
| Dilution | Neutralization 0.4-2 ug/mL |
| Applications | Neutralization |
Description
R-Spondin 2 Monoclonal specifically detects R-Spondin 2 in Human samples. It is validated for Neutralization.Specifications
| R-Spondin 2 | |
| LYOPH | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Human | |
| Q6UXX9 | |
| 340419 | |
| Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2 Met1-Gly205 Accession # Q6UXX9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human R-Spondin 2 in direct ELISAs. |
| 879709 | |
| Neutralization 0.4-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cristin 2, CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, RSPO2, RSpondin 2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 | |
| RSPO2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title