Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human R-Spondin 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$214.00 - $579.52
Specifications
| Antigen | R-Spondin 2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
R-Spondin 2 Polyclonal specifically detects R-Spondin 2 in Human samples. It is validated for Western Blot.Specifications
| R-Spondin 2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| Cristin 2, CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, RSPO2, RSpondin 2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 | |
| RSPO2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q6UXX9 | |
| 340419 | |
| E. coli-derived recombinant human R-Spondin 2 isoform 1 Cys21-Gly205 Accession # Q6UXX9 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human R-Spondin 2 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human (rh) R-Spondin 1, rhR-Spondin 3, and recombinant mouse R-Spondin 4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title