Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human R-Spondin 3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | R-Spondin 3 |
---|---|
Concentration | LYOPH |
Dilution | Neutralization 0.3-1.2 ug/mL |
Applications | Neutralization |
Classification | Polyclonal |
Description
R-Spondin 3 Polyclonal specifically detects R-Spondin 3 in Human samples. It is validated for Neutralization.Specifications
R-Spondin 3 | |
Neutralization 0.3-1.2 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2 | |
RSPO3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Neutralization | |
Unconjugated | |
RUO | |
Q9BXY4 | |
84870 | |
Chinese hamster ovary cell line CHO-derived recombinant human R-Spondin 3 Gln22-His272 Accession # Q9BXY4 | |
Primary | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Detects human R-Spondin 3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse R-Spondin 3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title