Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAB11FIP3 Partial ORF (NP_055515.1, 655 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098230
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]

Sequence: HSRARESELEQEVRRLKQDNRNLKEQNEELNGQIITLSIQGAKSLFSTAFSESLAAEISSVSRDELMEAIQKQEEINFRLQDYIDRIIVAIMETNPSILE

Specifications

Accession Number NP_055515.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9727
Molecular Weight (g/mol) 36.74kDa
Name RAB11FIP3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen HSRARESELEQEVRRLKQDNRNLKEQNEELNGQIITLSIQGAKSLFSTAFSESLAAEISSVSRDELMEAIQKQEEINFRLQDYIDRIIVAIMETNPSILE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0665/Rab11-FIP3
Common Name RAB11FIP3
Gene Symbol RAB11FIP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.