Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Rab27a Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $472.87
Specifications
| Antigen | Rab27a |
|---|---|
| Clone | 2537A |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7245SP
![]() |
R&D Systems
MAB7245SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7245100
![]() |
R&D Systems
MAB7245100 |
100 μg |
Each for $472.87
|
|
|||||
Description
Rab27a Monoclonal specifically detects Rab27a in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| Rab27a | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| GS2, GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAM, RAMras-related protein Rab-27A | |
| E. coli-derived recombinant human RAB27a, Ser135-Ala218, Accession # P51159 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2537A | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P51159 | |
| 5873 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title