Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RAGE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11451SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11451SP 25 μg
MAB11451100 100 μg
MAB11451500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB11451SP Supplier R&D Systems Supplier No. MAB11451SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 12 publications

AGER Monoclonal specifically detects AGER in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen RAGE
Applications Western Blot, Blockade of Receptor-ligand Interaction, ELISA
Classification Monoclonal
Clone 176902
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15109
Gene Alias advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1
Gene Symbols AGER
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RAGE Gln24-Ala344 Accession # Q15109
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Test Specificity Detects human RAGE in ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse RAGE and recombinant rat RAGE is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.