Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAI1 Partial ORF (NP_109590.3, 1 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959294
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene is located within the Smith-Magenis syndrome region on chromosome 17. It is highly similar to its mouse counterpart and is expressed at high levels mainly in neuronal tissues. The protein encoded by this gene includes a polymorphic polyglutamine tract in the N-terminal domain. Expression of the mouse counterpart in neurons is induced by retinoic acid. This gene is associated with both the severity of the phenotype and the response to medication in schizophrenic patients. [provided by RefSeq]

Sequence: MQSFRERCGFHGKQQNYQQTSQETSRLENYRQPSQAGLSCDRQRLLAKDYYNPQPYPSYEGGAGTPSGTAAAVAADKYHRGSKALPTQQGLQGRPAFPGYG

Specifications

Accession Number NP_109590.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10743
Molecular Weight (g/mol) 36.85kDa
Name RAI1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MQSFRERCGFHGKQQNYQQTSQETSRLENYRQPSQAGLSCDRQRLLAKDYYNPQPYPSYEGGAGTPSGTAAAVAADKYHRGSKALPTQQGLQGRPAFPGYG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434A139/KIAA1820/MGC12824/SMCR/SMS
Common Name RAI1
Gene Symbol RAI1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.